Would you like to log in again?
Rat IGF-2 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0197
76237-146EA 687 USD InStock
76237-146
Rat IGF-2 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Rat IGF-2 AccuSignal ELISA Kit
  • Natural and recombinant rat IGF-2.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: A25-E91
Insulin-like growth factor II is also known as somatomedin A. IGF-2 is a member of the insulin family of polypeptide growth factors that is involved in development and growth. It is paternally expressed in the fetus and placenta. IGF-II is a mitogen for many cell types and an important modulator of muscle growth and differentiation. IGF-II gene is prevalently expressed during prenatal development and its gene activity is regulated by genomic imprinting, in that the allele inherited from the father is active and the allele inherited from the mother is inactive in most normal tissues. IGF-II appears to be induced by placental lactogen during prenatal development. It is a mediator of prolactin-induced alveologenesis; prolactin, IGF-2, and cyclin D1, all of which are overexpressed in breast cancers, are components of a developmental pathway in the mammary gland.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml rat IGF-2 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each rat IGF-2 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty