Would you like to log in again?
Human IL-13 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0237
76237-154EA 636 USD InStock
76237-154
Human IL-13 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Interleukin-13 AccuSignal ELISA Kit
  • Natural and recombinant human IL-13.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: G21-N132
Interleukin 13 (IL-13) is a protein that in humans is encoded by the IL13 gene. IL-13 is cytokine secreted by many cell types, but especially T helper type 2 (Th2) cells which is an important mediator of allergic inflammation and disease. IL-13 induces its effects through a multi-subunit receptor that includes the alpha chain of the IL-4 receptor (IL-4R alpha) and at least one of two known IL-13-specific binding chains. Most of the biological effects of IL-13, like those of IL-4, are linked to a single transcription factor, signal transducer and activator of transcription 6 (STAT6). The IL13 gene is clustered with IL3, IL5, IL4, and CSF2 on 5q.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.3pg/ml, 15.6pg/ml human IL-13 standard solutions into the pre-coated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. We recommend that each human IL-13 standard solution and each sample is measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty