Would you like to log in again?
Mouse MDC ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0256
76237-156EA 652 USD InStock
76237-156
Mouse MDC ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse MDC AccuSignal ELISA Kit
  • Natural and recombinant mouse MDC.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence:  G25-S92
Macrophage-derived chemokine(MDC), also called Chemokine, cc motif, ligand 22(CCL22) or Small inducible cytokine subfamily A, member 22(SCY22). MDC is a recently identified member of the CC chemokine family. It is not closely related to other chemokines, sharing most similarity with thymus- and activation-regulated chemokine(TARC), which contains 37% identical amino acids. In addition, MDC gene is mapped to chromosome 16q13, the same position reported for the TARC gene. MDC has the four-cysteine motif and other highly conserved residues characteristic of CC chemokines, but it shares<35% identity with any of the known chemokines. Recombinant MDC was expressed in Chinese hamster ovary cells and purified by heparin-Sepharose chromatography. MDC is highly expressed in macrophages and in monocyte-derived dendritic cells, but not in monocytes, natural killer cells, or several cell lines of epithelial, endothelial, or fibroblast origin. High expression was also detected in normal thymus and less expression in lung and spleen. MDC is thus a unique member of the CC chemokine family that may play a fundamental role in the function of dendritic cells, natural killer cells, and monocytes.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml mouse MDC standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse MDC standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty