Would you like to log in again?
Human CCL4 - MIP-1 beta ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0259
76237-162EA 636 USD InStock
76237-162
Human CCL4 - MIP-1 beta ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human CCL4 - MIP-1 beta AccuSignal ELISA Kit
  • Natural and recombinant human CCL4.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence:  A24-N92
Chemokine(C-C motif) ligand 4, also known as CCL4, is a protein which in humans is encoded by the CCL4 gene. It is a CC chemokine with specificity for CCR5 receptors. It is a chemoattractant for natural killer cells, monocytes and a variety of other immune cells. CCL4 is a major HIV-suppressive factor produced by CD8+ T cells. Performing-low memory CD8+ T cells that normally synthesize MIP-1-beta. the CCL4 gene was assigned to a slightly more distal location: 17q21-q23 rather than 17q11-q21.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.3pg/ml, 15.6pg/ml, 7.8pg/ml human CCL4 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA, citrate) to each empty well. It is recommended that each human CCL4 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty