Would you like to log in again?
Human PLGF ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0292
76237-164EA 658 USD InStock
76237-164
Human PLGF ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human PLGF AccuSignal ELISA Kit
  • Natural and recombinant human PLGF.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: A21-R149
The protein, also called PGF, is 149 amino acids long and shares 53% identity with the platelet-derived growth factor-like region of human VPF. And the N-glycosylated PLGF protein is secreted into the medium and that it functions as a dimer1. The PLGF gene is mapped to 14q24-q31. There are 3 isoforms of PGF, designated PGF1, PGF2, and PGF3. Only PGF2 is able to bind heparin. Additionally, PGF regulates inter- and intramolecular cross-talk between the VEGF receptor tyrosine kinases FLT1 and FLK12. It also can stimulate angiogenesis and collateral growth in ischemic heart and limb with at least a comparable efficiency to VEGF3.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml human PLGF standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma(heparin, EDTA, citrate) or urine to each empty well. It is recommended that each human PLGF standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty