Would you like to log in again?
Human P-Selectin ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0301
76237-180EA 636 USD InStock
76237-180
Human P-Selectin ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human P-Selectin AccuSignal ELISA Kit
  • Natural and recombinant human sP-Selectin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: W42-A771
P-selectin, also called GMP-140, CD62, or selectin P, is a 140-kD adhesion molecule, expressed at the surface of activated cells, that mediates the interaction of activated endothelial cells or platelets with leukocytes. It is stored in secretory granules and expressed at the plasma membrane after cell activation. It is known to play an important role in atherosclerosis. The major ligand for P-selectin on leukocytes is P-selectin glycoprotein ligand-1(PSGL-1). The standard product used in this kit is recombinant human P-Selectin, excluding intercellular P-Selectin and transmembrane domain. It has 730 amino acids sequence with the molecular mass of 80 KDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human P-Selectin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA, citrate) to each empty well. It is recommended that each human P-Selectin standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty