Would you like to log in again?
Human IL-20 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0367
76237-200EA 636 USD InStock
76237-200
Human IL-20 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Interleukin-20 AccuSignal ELISA Kit
  • Natural and recombinant human IL-20.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: L25-E176
Interleukin-20(IL-20) is a protein belonging to the IL-10 family of cytokines. IL-20 is produced by activated keratinocytes and monocytes and transmits an intracellular signal through two distinct cell-surface receptor complexes on keratinocytes and other epithelial cells. By radiation hybrid analysis, the IL20 gene was mapped to chromosome 1q32, where it is tightly linked to the IL10, IL19, and MDA7 genes within a 195-kb region, the IL10 family cytokine cluster. IL-20 regulates proliferation and differentiation of keratinocytes during inflammation, particularly inflammation associated with the skin. In addition, IL-20 also causes cell expansion of multipotential hematopoietic progenitor cells.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human IL-20 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human IL-20 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty