Would you like to log in again?
Human CD105 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0371
76237-208EA 687 USD InStock
76237-208
Human CD105 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human CD105 AccuSignal ELISA Kit
  • Natural and recombinant human CD105.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: CHO; Immunogen sequence: E26-G586
CD105, also called Endoglin, is a homodimeric membrane glycoprotein primarily associated with human vascular endothelium. It is also found on bone marrow proerythroblasts, activated monocytes, and lymphoblasts in childhood leukemia. Endoglin is a component of the transforming growth factor-beta(TGFB) receptor complex and binds TGFB1 with high affinity.1 CD105 gene is mapped to 9q34.1. The coding region of the gene contains 14 exons.2 The protein consists of a homodimer of 180 kDA with disulfide links. Endoglin has a role in the development of the cardiovascular system and in vascular remodeling. Its expression is regulated during heart development. Furthermore, it also has a role in the balance of ALK1 and ALK5 signaling to regulate endothelial cell proliferation.3 Moreover, the elevated expression of endoglin in the surgically excised CNVMs suggested a persisting postmitotic activation in an advanced stage of neovascular tissue.4 The standard product used in this kit is extracellular part of recombinant huma...
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human CD105 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human CD105 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty