Would you like to log in again?
Human Angiopoietin-2 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0374
76237-214EA 636 USD InStock
76237-214
Human Angiopoietin-2 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Angiopoietin-2 AccuSignal ELISA Kit
  • Natural and recombinant human Angiopoietin-2.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: Y19-F496
Angiopoietin 1 and Angiopoietin 2 are important for development of the endothelium, by regulating tyrosine phosphorylation of the membrane receptor Tie 2. Angiopoietin 2 is only 60% homologous with Angiopoietin 1. Angiopoietin-2 is a naturally occurring antagonist of angiopoietin-1 that competes for binding to the TIE2 receptor and blocks ANGPT1-induced TIE2 autophosphorylation. Angiopoietin 1 binding to Tie 2 causes phosphorylation of the receptor. Angiopoietin 2 competes for this binding, and thus blocks receptor phosphorylation. Angiopoietin 2 expression occurs at sites of vascular remodelling: dorsal aorta and major aortic branches, ovary, placenta and uterus.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human Angiopoietin-2 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma(heparin, EDTA), saliva or human milk to each empty well. It is recommended that each human Angiopoietin-2 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty