Would you like to log in again?
Mouse BAFF ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0381
76237-228EA 686.86 USD InStock
76237-228
Mouse BAFF ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse BAFF AccuSignal ELISA Kit
  • Natural and recombinant mouse BAFF.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: A127-L309
B-cell activating factor(BAFF) also known as tumor necrosis factor ligand superfamily member 13B is a protein that in humans is encoded by the TNFLSF13B gene.1,2 BAFF is a cytokine that belongs to the tumor necrosis factor(TNF) ligand family. This cytokine is a ligand for receptors TNFRSF13B/TACI, TNFRSF17/BCMA, and TNFRSF13C/BAFFR. This cytokine is expressed in B cell lineage cells, and acts as a potent B cell activator. It has been also shown to play an important role in the proliferation and differentiation of B cells.3All these ligands act as heterotrimers(i.e. three of the same molecule) interacting with heterotrimeric receptors4, although BAFF has been known to be active as either a hetero- or homotrimer.5 The standard used in this kit is recombinant mouse soluble BAFF(A127-L309) with molecular weight 23.2KDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 3000pg/ml, 1500pg/ml, 750pg/ml, 419pg/ml, 188pg/ml, 94pg/ml, 46.9pg/ml mouse BAFF standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. We recommend that each mouse BAFF standard solution and each sample is measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty