Would you like to log in again?
Mouse P-Cadherin ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0383
76237-232EA 636 USD InStock
76237-232
Mouse P-Cadherin ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse P-Cadherin AccuSignal ELISA Kit
  • Oligomeric forms of natural and recombinant mouse P-Cadherin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: E100-G647
P-Cadherin, also known as Cadherin-3(CDH3), is a protein that in humans is encoded by the CDH3 gene. This gene is a classical cadherin from the cadherin superfamily. This gene is located in a six-cadherin cluster in a region on the long arm of chromosome 16 that is involved in loss of heterozygosity events in breast and prostate cancer. In addition, aberrant expression of this protein is observed in cervical adenocarcinomas. Mutations in this gene have been associated with congential hypotrichosis with juvenile macular dystrophy. Cells expressing CDH3 also adhered to one another more tightly than the parental cell line.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml mouse P-Cadherin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates or serum to each empty well. It is recommended that each mouse P-Cadherin standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty