Would you like to log in again?
Human CCL28 EKISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0398
76237-254EA 636 USD InStock
76237-254
Human CCL28 EKISA Kit, Rockland Immunochemicals
Assays ELISAs
Human CCL28 EKISA Kit
  • Natural and recombinant human CCL28.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: I23-Y127
CCL28, also known as mucosae-associated epithelial chemokine (MEC), CCK1 and SCYA28, is a chemokine. It is mapped to 5p12. The tumor hypoxia promotes the recruitment of regulatory T (Treg) cells through induction of expression of the chemokine CCL28, which in turn promotes tumor tolerance and angiogenesis. CCL28 regulates the chemotaxis of cells that express the chemokine receptors CCR3 and CCR10. CCL28 has also been implicated in the migration of IgA-expressing cells to the mammary gland, salivary gland, intestine and other mucosal tissues. It has also been shown as a potential antimicrobial agent effective against certain pathogens, such as Gram negative and Gram positive bacteria and the fungus Candida albicans.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml human CCL28 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum and plasma(heparin, EDTA) to each empty well. It is recommended that each human CCL28 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty