Would you like to log in again?
Mouse CD14 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0400
76237-258EA 636 USD InStock
76237-258
Mouse CD14 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse CD14 AccuSignal ELISA Kit
  • Natural and recombinant mouse CD14.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: A18-P345
CD14, Cluster of differentiation 14, ingle-copy gene encoding 2 protein forms: a 50- to 55-kD glycosylphosphatidylinositol-anchored membrane protein(mCD14) and a monocyte or liver-derived soluble serum protein(sCD14) that lacks the anchor. By in situ hybridization and study of somatic cell hybrid DNA that the gene is located at bands 5q23-q31. CD14 acts as a co-receptor(along with the Toll-like receptor TLR 4 and MD-2) for the detection of bacterial lipopolysaccharide(LPS). CD14 can bind LPS only in the presence of lipopolysaccharide-binding protein(LBP). Although LPS is considered its main ligand, CD14 also recognizes other pathogen-associated molecular patterns.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml mouse CD14 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse CD14 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty