Would you like to log in again?
Mouse CD36 - SR-B3 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0406
76237-270EA 636 USD InStock
76237-270
Mouse CD36 - SR-B3 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse CD36 - SR-B3 AccuSignal ELISA Kit
  • Natural and recombinant mouse CD36.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: CHO; Immunogen sequence: G30-K439
CD36(Cluster of Differentiation 36) is an integral membrane protein found on the surface of many cell types in vertebrate animals and is also known as FAT, SCARB3, GP88, glycoprotein IV(gpIV) and glycoprotein IIIb(gpIIIb). The human CD36 is a member of a gene family of structurally related glycoproteins and functions as a receptor for collagen type I and thrombospondin. The use of a CD36 genomic probe has allowed the localization of the CD36 locus to the 7q11.2 band by fluorescence in situ hybridization coupled with GTG-banding. CED-1/SCARF1 and C03F11.3/CD36 are beta-glucan binding receptors and define an evolutionarily conserved pathway for the innate sensing of fungal pathogens.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml mouse CD36 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse CD36 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty