Would you like to log in again?
Human CD40 - TNFRSF5 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0407
76237-272EA 687 USD InStock
76237-272
Human CD40 - TNFRSF5 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human CD40 - TNFRSF5 AccuSignal ELISA Kit
  • Natural and recombinant human CD40.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: E21-R193
CD40 is a cell surface receptor that is expressed on the surface of all mature B cells, most mature B-cell malignancies, and some early B-cell acute lymphocytic leukemias, but is not expressed on plasma cells. The CD40 gene is localized to the long arm of chromosome 20, bands q12-q13.2. The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor has been found to be essential in mediating a broad variety of immune and inflammatory responses including T cell-dependent immunoglobulin class switching, memory B cell development, and germinal center formation.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml human CD40 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma( heparin, EDTA), saliva, urine or human milk to each empty well. It is recommended that each human CD40 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty