Would you like to log in again?
Rat CXCL1 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0421
76237-284EA 687 USD InStock
76237-284
Rat CXCL1 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Rat CXCL1 AccuSignal ELISA Kit
  • Natural and recombinant rat CXCL1.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: A25-K96
Chemokine(C-X-C motif) ligand 1(CXCL1) is a small cytokine belonging to the CXC chemokine family that was previously called GRO1 oncogene, GROalpha, KC, Neutrophil-activating protein 3(NAP-3) and melanoma growth stimulating activity, alpha(MSGA-alpha). In humans, this protein is encoded by the CXCL1 gene. The gene for CXCL1 is located on human chromosome 4 amongst genes for other CXC chemokines. The mature form of CXCL1 is maximally 73 amino acids long. CXCL1 is secreted by human melanoma cells, has mitogenic properties and is implicated in melanoma pathogenesis. CXCL1 is expressed by macrophages, neutrophils and epithelial cells, and has neutrophil chemoattractant activity. This chemokine elicits its effects by signaling through the chemokine receptor CXCR2.CXCL1 decreased the severity of multiple sclerosis and may offer a neuro-protective function.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml rat CXCL1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernatants, serum or plasma(EDTA) to each empty well. It is recommended that each rat CXCL1 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty