Would you like to log in again?
Rat CXCL2 - MIP-2 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0422
76237-286EA 636 USD InStock
76237-286
Rat CXCL2 - MIP-2 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Rat CXCL2 - MIP-2 AccuSignal ELISA Kit
  • Natural and recombinant rat MIP-2.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: S32-N100
MIP is a member of the aquaporin family of membrane-bound water channels. MIP family proteins are thought to contain 6 TM domains. Sequence analysis suggests that the proteins may have arisen through tandem, intragenic duplication from an ancestral protein that contained 3 TM domains. Major intrinsic protein (MIP, also called MP26) is the predominant fiber cell membrane protein of the ocular lens. The major intrinsic protein (MIP) of the vertebrate eye lens is the first identified member of a sequence-related family of cell-membrane proteins that appears to have evolved by gene duplication. Several members of the MIP family transport water (aquaporins), glycerol and other small molecules in microbial, plant and animal cells.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml rat MIP-2 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each rat MIP-2 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty