Would you like to log in again?
Human IL-17E - IL-25 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0468
76237-322EA 687 USD InStock
76237-322
Human IL-17E - IL-25 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Interleukin-17E - Interleukin-25 AccuSignal ELISA Kit
  • Natural and recombinant human IL-17E.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.colI; Immunogen sequence: Y33-G177
Interleukin-25 (IL-25), also known as interleukin-17E (IL-17E), is a protein that in humans is encoded by the IL25 gene. It is a cytokine that belongs to the IL-17 cytokine family and mapped to 14q11.2. IL-25 is secreted by type 2 helper T cells (Th2) and mast cells. It has been found that IL-25 can induce Th2-type cytokine expression and production by non-T, non-B accessory cells that express high levels of major histocompatibility complex class II and low levels of Cd11c. This cytokine also can induce NF-kB activation, and stimulate the production of IL8. What’s more, IL-25 is an important molecule controlling immunity of the gut and has been implicated in chronic inflammation associated with the gastrointestinal tract.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human IL-17E standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. We recommend that each human IL-17E standard solution and each sample is measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty