Would you like to log in again?
Human IL-27 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0472
76237-328EA 736 USD InStock
76237-328
Human IL-27 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Interleukin-27 AccuSignal ELISA Kit
  • Natural and recombinant human IL-27.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: EBI3(R21-K229)+P28(F29-P243)
Interleukin-27(IL-27) is a heterodimeric cytokine belonging to the IL-12 family that is composed of two subunits; Epstein-Barr virus(EBV)-induced gene 3(EBI3)(also known as IL-27B) and IL27-p28(known as IL-30). IL-27 is produced by antigen-presenting cells.IL-27 plays an important function in regulating the activity of B- and T-lymphocytes. IL-27, a potent inhibitor of T(H)-17 cell development, may be a useful target for treating inflammatory diseases mediated by these cells.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human IL-27 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human IL-27 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty