Would you like to log in again?
Human LOX-1 - OLR1 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0492
76237-344EA 636 USD InStock
76237-344
Human LOX-1 - OLR1 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human LOX-1 - OLR1 AccuSignal ELISA Kit
  • Natural and recombinant human LOX-1.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: S61-Q273
LOX-1(lectin-type oxidized LDL receptor 1) also known as OLR1, is a protein that in humans is encoded by the OLR1 gene. LOX-1 is a receptor protein which belongs to the C-type lectin superfamily. The LOX1 gene is mapped to 12p13-p12 by fluorescence in situ hybridization. LOX1 is expressed on the plasma membrane of differentiated macrophages, but not on monocytes. The LOX1 protein acts as a macrophage scavenger receptor. LOX1 expression was detected in all choroidal neovascular membranes, regardless of structure, whereas there was no evidence of LOX1 within the posterior segments of normal eyes. LOX1 plays an active role in the pathogenesis of choroidal neovascularization, especially in ARMD.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml human LOX-1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum and plasma(heparin, EDTA) to each empty well. It is recommended that each human LOX-1 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty