Would you like to log in again?
Mouse LOX-1 - OLR1 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0493
76237-346EA 713 USD InStock
76237-346
Mouse LOX-1 - OLR1 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse LOX-1 - OLR1 AccuSignal ELISA Kit
  • Natural and recombinant mouse LOX-1.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: R60-I363
Oxidized low-density lipoprotein receptor 1(Ox-LDL receptor 1) also known as lectin-type oxidized LDL receptor 1(LOX-1) is a protein that in humans is encoded by the OLR1 gene. It belongs to the C-type lectin superfamily. The LOX1 gene is mapped to to 12p13-p12. The protein has got the amino acids of 363. It can be detected in various tissues, highly expressed in placenta. This protein may be involved in the regulation of Fas-induced apoptosis and play a role as a scavenger receptor. The standards of this kit are recombinant mouse LOX-1(R60-I363), with molecular weight of 36.3kDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml mouse LOX-1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse LOX-1 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty