Would you like to log in again?
Mouse RAGE ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0495
76237-350EA 636 USD InStock
76237-350
Mouse RAGE ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse RAGE AccuSignal ELISA Kit
  • Natural and recombinant mouse RAGE.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: Q24-A342
RAGE, the Receptor for Advanced Glycation Endproducts, is a 35kD transmembrane receptor of the immunoglobulin super family. It is also known as “AGER”. AGER gene is mapped to chromosome 6p21.3 by mapping by contiguous cosmids and YAC clones and by fluorescence in situ hybridization. The expression of RAGE is particularly increased in neurons close to deposits of amyloid beta peptide and to neurofibrillary tangles. RAGE has been linked to several chronic diseases, which are thought to result from vascular damage. The pathogenesis is hypothesized to include ligand binding upon which RAGE signals activation of the nuclear factor kappa B(NF-kappaB).
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml, 78pg/ml mouse RAGE standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernatants, serum or plasma(heparin, EDTA) to each empty well. We recommend that each mouse RAGE standard solution and each sample is measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty