Would you like to log in again?
Human RBP4 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0498
76237-356EA 687 USD InStock
76237-356
Human RBP4 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human RBP4 AccuSignal ELISA Kit
  • Natural and recombinant human RBP4.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: E19-L201
Retinol binding protein 4, plasma, also known as RBP4, belongs to the lipocalin family and is the specific carrier for retinol(vitamin A alcohol) in the blood. It is protein that in humans is encoded by the RBP4 gene. RBP4 gene resides just centromeric of the cluster of CYP2C genes on 10q24. The mouse Rbp4 locus is closely linked and just proximal to the locus for phenobarbital-inducible cytochrome P450-2c(Cyp-2c) at the distal end of chromosome 19. It delivers retinol from the liver stores to the peripheral tissues. In plasma, the RBP-retinol complex interacts with transthyretin, which prevents its loss by filtration through the kidney glomeruli. A deficiency of vitamin A blocks secretion of the binding protein posttranslationally and results in defective delivery and supply to the epidermal cells. The standard used in this kit is recombinant protein, with E19-L201 aa sequence, the molecular weight is 22kda.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 20000pg/ml, 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml human RBP4 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma (heparin, EDTA) or urine to each empty well. We recommend that each human RBP4 standard solution and each sample is measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty