Would you like to log in again?
Human Kallistatin - Serpina4 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0502
76237-364EA 687 USD InStock
76237-364
Human Kallistatin - Serpina4 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Kallistatin - Serpina4 AccuSignal ELISA Kit
  • Natural and recombinant human Kallistatin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: Q21-P427
Serpin peptidase inhibitor, clade A(alpha-1 antiproteinase, antitrypsin), member 4, also called Kallistatin, is a protein that in humans is encoded by the SERPINA4 gene. This gene is mapped to chromosome 14q32.13. Kallistatin is a serine proteinase inhibitor(serpin) and a heparin-binding protein. It is localized in vascular smooth muscle cells and endothelial cells of blood vessels, suggesting that it may be involved in the regulation of vascular function. It shows that kallistatin plays a role in neointima hyperplasia. The potential role of kallistatin was investigated in angiogenesis in vitro and in vivo, and presented results demonstrating a novel role of the protein in the inhibition of angiogenesis and tumor growth.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human Kallistatin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human Kallistatin standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty