Would you like to log in again?
Human TLR3 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0547
76237-390EA 687 USD InStock
76237-390
Human TLR3 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human TLR3 AccuSignal ELISA Kit
  • Natural and recombinant human TLR3.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: K27-S711
Toll-like receptor 3(TLR3) also known as CD283(cluster of differentiation 283) is a protein that in humans is encoded by the TLR3 gene. The TLR3 gene was mapped to chromosome 4q35 by fluorescence in situ hybridization. TLR3 specifically recognizes double-stranded RNA(dsRNA) and induces multiple intracellular events responsible for innate antiviral immunity against a number of viral infections.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human TLR3 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human TLR3 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty