Would you like to log in again?
Mouse CD25 - IL-2sR alpha ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0552
76237-400EA 636 USD InStock
76237-400
Mouse CD25 - IL-2sR alpha ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse CD25 - Interleukin-2sR alpha AccuSignal ELISA Kit
  • Natural and recombinant mouse CD25.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: E22-K236
CD25 is the alpha chain of the IL-2 receptor. It is a type I transmembrane protein present on activated T cells, activated B cells, some thymocytes, myeloid precursors, and oligodendrocytes that associates with CD122 to form a heterodimer that can act as a high-affinity receptor for IL-2. CD25 has been used as a marker to identify CD4+FoxP3+ regulatory T cells in mice. By in situ hybridization, the IL2R gene was mapped to 10p15-p14. Its soluble form, called sIL-2R may be elevated in these diseases and is occasionally used to track disease progression.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 313pg/ml, 156pg/ml, 78pg/ml mouse CD25 standard solutions into the pre-coated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma( heparin, EDTA) to each empty well. We recommend that each mouse CD25 standard solution and each sample is measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty