Would you like to log in again?
Human ADAMTS13 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KOA0563
76237-414EA 687 USD InStock
76237-414
Human ADAMTS13 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human ADAMTS13 AccuSignal ELISA Kit
  • Natural and recombinant human ADAMTS13.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: CHO; Immunogen sequence: Q34-W688
ADAMTS13 is a zinc-containing metalloprotease enzyme. The ADAMTS13 gene is mapped to chromosome 9q34 by genomic sequence analysis. It is reported that a metal-containing proteolytic enzyme (metalloprotease) in normal plasma cleaves the peptide bond between tyrosine at position 842 and methionine at position 843 in monomeric subunits of von Willebrand factor, thereby degrading the large multimers. And it is confirmed that the ADAMTS13 gene encodes the von Willebrand factor-cleaving protease (VWFCP). ADAMTS13 is secreted in blood and degrades large vWf multimers, decreasing their activity.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 50ng/ml, 25ng/ml, 12.5ng/ml, 6.25ng/ml, 3.12ng/ml, 1.56ng/ml, 0.78ng/ml human ADAMTS13 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA, citrate) to each empty well. We recommend that each human ADAMTS13 standard solution and each sample is measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty