Would you like to log in again?
Human Tissue factor - F3 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0564
76237-416EA 636 USD InStock
76237-416
Human Tissue factor - F3 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Tissue factor - F3 AccuSignal ELISA Kit
  • Natural and recombinant human Tissue factor.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: S33-E251
Tissue factor, also called platelet tissue factor, factor III, thrombokinase, or CD142 is a protein present in sub endothelial tissue and leukocytes necessary for the initiation of thrombin formation from the zymogene prothrombin. An incorrect synonym is thromboplastin. the F3 gene was mapped to 1pter-p21 by study of somatic cell hybrids with a species-specific sensitive chromogenic assay. This gene encodes coagulation factor III which is a cell surface glycoprotein. This factor enables cells to initiate the blood coagulation cascades, and it functions as the high-affinity receptor for the coagulation factor VII.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml human Tissue factor standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma(heparin, EDTA, citrate) cell lysates or urine to each empty well. It is recommended that each human Tissue factor standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty