Would you like to log in again?
Mouse IL-33 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0566
76237-420EA 636 USD InStock
76237-420
Mouse IL-33 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse Interleukin-33 AccuSignal ELISA Kit
  • Natural and recombinant mouse IL-33.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: S109-I266
Interleukin 33(IL-33) is a cytokine belonging to the IL-1 superfamily. By genomic sequence analysis, the IL33 gene is mapped to chromosome 9p24.1 and its mouse homolog to a syntenic region on chromosome 19qC1. The induction of type 2 cytokines by IL-33 in vivo is believed to induce the severe pathological changes observed in mucosal organs following administration of IL-33. IL33, an alarmin released from necrotic cells, is necessary for potent CD8 + T cell(CTL) responses to replicating, prototypic RNA, and DNA viruses in mice. IL33 prevented the downregulation of CXCR2 and inhibition of chemotaxis induced by activation of TLR4, and found that IL33 reverses the TLR4-induced reduction of CXCR2 expression via the inhibition of expression of GRK2.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml mouse IL-33 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse IL-33 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty