Would you like to log in again?
Human ADAM12 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0569
76237-426EA 636 USD InStock
76237-426
Human ADAM12 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human ADAM12 AccuSignal ELISA Kit
  • Natural and recombinant human ADAM12.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: CHO; Immunogen sequence: R29-S513
ADAM12(Disintegrin and metalloproteinase domain-containing protein 12), also known as MLTN, is an enzyme that in humans is encoded by the ADAM12 gene. This gene encodes a member of the ADAM(a disintegrin and metalloprotease) protein family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This gene has two alternatively spliced transcripts: a shorter secreted form and a longer membrane-bound form. The shorter form is found to stimulate myogenesis. By RT-PCR and immunoblot analyses that expression of mouse Adam12 increases during muscle regeneration, while the levels of other ADAMs remain constant. Immunofluorescence analysis revealed staining of small, newly formed muscle fibers in regenerating but not normal adult muscle cells. By using of fluorescence in situ hybridization, the ADAM12 gene is mapped to human chromosome 10q26.3.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human ADAM12 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma(heparin) or urine to each empty well. It is recom
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty