Would you like to log in again?
Mouse TEK - TIE2 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0604
76237-438EA 636 USD InStock
76237-438
Mouse TEK - TIE2 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse TEK - TIE2 AccuSignal ELISA Kit
  • Natural and recombinant mouse TEK.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: A23-K744
Angiopoietin-1 receptor, also called TEK or TIE2 is a protein that in humans is encoded by the TEK gene. This gene is mapped to 9p21.2. The TEK receptor tyrosine kinase is expressed almost exclusively in endothelial cells in mice, rats, and humans. This receptor possesses a unique extracellular domain containing 2 immunoglobulin-like loops separated by 3 epidermal growth factor-like repeats that are connected to 3 fibronectin type III-like repeats. The ligand for the receptor is angiopoietin-1. Defects in TEK are associated with inherited venous malformations, the TEK signaling pathway appears to be critical for endothelial cell-smooth muscle cell communication in venous morphogenesis. TEK is closely related to the TIE receptor tyrosine kinase.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 8000pg/ml, 4000pg/ml, 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml mouse TEK standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum, tissue homogenates or plasma (heparin, EDTA) to each empty well. It is recommended that each mouse TEK standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty