Would you like to log in again?
Human TGF-beta 2 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0609
76237-448EA 636 USD InStock
76237-448
Human TGF-beta 2 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human TGF-beta 2 AccuSignal ELISA Kit
  • Natural and recombinant human TGF-beta 2.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: A303-S414
Transforming growth factor-beta 2(TGF-beta 2) is a secreted protein known as a cytokine that performs many cellular functions and has a vital role during embryonic development. This gene is mapped to 1q41. It is an extracellular glycosylated protein. It is known to suppress the effects of interleukin dependent T-cell tumors. TGF-beta 2 is present at elevated levels in the aqueous humor of patients with primary open angle glaucoma(POAG). Studies have shown that TGF-beta 2 influences cultured trabecular meshwork cells, and it reduced outflow facility when perfused into cultured human anterior segments. In POAG, elevated expression of Gremlin by TM cells inhibited BMP4 antagonism of TGF-beta 2 and led to increased extracellular matrix deposition and elevated IOP.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml human TGF-beta 2 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of activated human cell culture supernates, serum or plasma(heparin, EDTA, citrate) to each empty well. It is recommended that each human TGF-beta 2 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty