Would you like to log in again?
Human CCL14 - HCC-1 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0643
76237-452EA 636 USD InStock
76237-452
Human CCL14 - HCC-1 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human CCL14 - HCC-1 AccuSignal ELISA Kit
  • Natural and recombinant human CCL14.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: T20-N93
Chemokine(C-C motif) ligand 14(CCL14) is a small cytokine belonging to the CC chemokine family. It is also commonly known as HCC-1. It is produced as a protein precursor that is processed to generate a mature active protein containing 74 amino acids that and is 46% identical in amino acid composition to CCL3 and CCL4. This chemokine is expressed in various tissues including spleen, bone marrow, liver, muscle, and gut. CCL14 activates monocytes, but does not induce their chemotaxis. In addition, HCC-1 enhanced the proliferation of CD34+ myeloid progenitor cells. It was as effective as MIP-1 alpha, but about 100-fold less potent. Human CCL14 is located on chromosome 17 within a cluster of other chemokines belonging to the CC family.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.3pg/ml, 15.6pg/ml human CCL14 standard solutions into the pre-coated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. We recommend that each human CCL14 standard solution and each sample is measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty