Would you like to log in again?
Rat MMP-8 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0734
76237-464EA 687 USD InStock
76237-464
Rat MMP-8 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Rat MMP-8 AccuSignal ELISA Kit
  • Natural and recombinant rat MMP-8.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: L21-P466
Matrix metalloproteinase 8(MMP8) also called neutrophil collagenase. Neutrophil collagenase, a member of the family of matrix metalloproteinases, is distinct from the collagenase of skin fibroblasts and synovial cells in substrate specificity and immunologic crossreactivity. MMP8, an enzyme that degrades fibrillar collagens imparting strength to the fetal membranes, is expressed by leukocytes and chorionic cytotrophoblast cells. The human neutrophil collagenase(HNC) cDNA clone has been sequenced and shown to encode a 467-residue protein. Neutrophil collagenase has been found to possess 57% identity with the deduced protein sequence for fibroblast collagenase with 72% chemical similarity. Certain regions of the molecule, including the putative zinc-binding region, are highly conserved. When compared with the published sequence for fibroblast collagenase, neutrophil collagenase contains four additional sites for glycosylation. The standard product used in this kit is natural, isolating from human MMP-8. The detected MMP-8 includes zymogen and active enzyme.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml rat MMP-8 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernates, serum or plasma(heparin) to each empty well. It is recommended that each rat MMP-8 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty