Would you like to log in again?
Rat CCL7 - MCP-3 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0774
76237-488EA 636 USD InStock
76237-488
Rat CCL7 - MCP-3 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Rat CCL7 - MCP-3 AccuSignal ELISA Kit
  • Natural and recombinant rat CCL7.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: Q24-P97
Chemokine(C-C motif) ligand 7(CCL7) is a small cytokine known as a chemokine that was previously called monocyte-specific chemokine 3(MCP3). It belongs to the C-C chemokine family. By fluorescence in situ hybridization, mapped the MCP3 gene to chromosome 17q11.2-q12. MCP3 was identified as a physiologic substrate of gelatinase A. Cleaved MCP3 binds to CC-chemokine receptors-1, -2, and -3, but no longer induces calcium fluxes or promotes chemotaxis, and instead acts as a general chemokine antagonist that dampens inflammation, suggested that matrix metalloproteinases are both effectors and regulators of the inflammatory response.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml rat CCL7 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each rat CCL7 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty