Would you like to log in again?
Rat Neuropilin-2 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0810
76237-496EA 636 USD InStock
76237-496
Rat Neuropilin-2 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Rat Neuropilin-2 AccuSignal ELISA Kit
  • Natural and recombinant rat Neuropilin-2.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: Q23-D857
NRP2(Neuropilin-2), also called Npn2 or VEGF165R2, encodes a member of the neuropilin family of receptor proteins. The soluble NRP2 was secreted as a 62.5-kD protein following transfection in Chinese hamster ovary cells. The NRP2 gene is mapped to 2q33.3. The NRP2 gene contains 17 exons and spans about 112 kb. This protein may play a role in cardiovascular development, axon guidance, and tumorigenesis. Mice with null mutations in genes encoding Sema3F, and its holoreceptor components Npn2 and plexin A3 (PLEXA3), exhibit increased dentate gyrus granule cell and cortical layer V pyramidal neuron spine number and size, and also aberrant spine distribution. Moreover, Sema3F promotes loss of spines and excitatory synapses in dissociated neurons in vitro, and in NRP2-null brain slices cortical layer V and dentate gyrus granule cells exhibit increased miniature excitatory postsynaptic current frequency. These disparate effects of secreted semaphorins are reflected in the restricted dendritic localization of NRP2 to ...
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml rat Neuropilin-2 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernates, cell lysates or tissue homogenates to each empty well. It is recommended that each rat Neuropilin-2 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty