Would you like to log in again?
AccuSignal DNA Damage (8-OHdG) ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0887
76237-546EA 663 USD InStock
76237-546
AccuSignal DNA Damage (8-OHdG) ELISA Kit, Rockland Immunochemicals
Assays ELISAs
AccuSignal DNA Damage 8-hydroxy-2-deoxy Guanosine (8-OHdG) ELISA Kit
  • Colorimetric Competitive ELISA (Enzyme-linked Immunosorbent Assay)
  • Sensitivity: 0.59 ng/mL. Range: 0.94-60 ng/mL.
8-hydroxy-2-deoxy Guanosine (8-OHdG) is produced by the oxidative damage of DNA by reactive oxygen and nitrogen species and serves as an established marker of oxidative stress. Hydroxylation of guanosine occurs in response to both normal metabolic processes and a variety of environmental factors (i.e., anything that increases reactive oxygen and nitrogen species). Increased levels of 8-OHdG are associated with the aging process as well as with a number of pathological conditions including cancer, diabetes, and hypertension. In complex samples such as plasma, cell lysates, and tissues, 8-OHdG can exist as either the free nucleoside or incorporated in DNA. Once the blood enters the kidney, free 8-OHdG is readily filtered into the urine, while larger DNA fragments remain in the bloodstream. Because of the complexity of plasma samples, urine is a more suitable matrix for the measurement of free 8-OHdG than plasma. Urinary levels of 8-OHdG range between 2.7-13 ng/mg creatine, while plasma levels of free 8-OHdG have been reported to be between 4-21 pg/ml as determined by LC-MS.
Colorimetric Competitive ELISA (Enzyme-linked Immunosorbent Assay) assay for quantification of 8-hydroxy-2-deoxy Guanosine in Cell lysates, Plasma, Sample Matrices, and Urine. Sensitivity: 0.59 ng/mL. Range: 0.94-60 ng/mL.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty