Would you like to log in again?
Normal Saline
Supplier: Rockland Immunochemicals
MB-049-1000
76237-548EA 81.71 USD InStock
76237-548
Normal Saline
Buffers Biological Buffers
This solution is commonly referred to as physiological saline or isotonic saline.
  • 0.15 M Sodium Chloride
  • Aseptically filtered through a Millipore 0.22 micron filter into clean, pre-sterilized containers
  • Tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria
Application Note: Saline solutions are available in various formulations for different purposes. Salines are also used in cell biology, molecular biology, and biochemistry experiments where ever normal salines are needed.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty