Would you like to log in again?
Mouse ICAM-1 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0188
76237-580EA 687 USD InStock
76237-580
Mouse ICAM-1 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse ICAM-1 AccuSignal ELISA Kit
  • Natural and recombinant mouse sICAM-1.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: Q28-N485
Intercellular adhesion molecule-1(ICAM-1) is an integral membrane protein, a member of the immunoglobulin superfamily, and a ligand for lymphocyte function-associated(LFA) antigens, a beta 2 leukocyte integrin. The normal function of human intercellular adhesion molecule-1(ICAM-1) is to provide adhesion between endothelial cells and leukocytes after injury or stress. ICAM-1 binds to leukocyte function-associated antigen(LFA-1) or macrophage-1 antigen(Mac-1). ICAM-1 is found on leukocytes, fibroblasts, epithelial cells and endothelial cells and its expression is regulated by inflammatory cytokines. ICAM-1 has a tissue distribution similar to that of the major histocompatibility complex class II antigens and is likely to play a role in inflammatory responses. The standard product used in this kit is produced by recombinant gene expression, which is a dimeric protein linked with disulfide bonds. The single chain is Gln28-Asn 485. It has 457 amino acids with the molecular mass of 76.8kDa. As a result of glycosylation, the molecular mass of 110-130KDa is revealed by SDS-PAGE.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml mouse ICAM-1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernatants, serum, plasma(heparin, EDTA) or tissue lysates to each empty well. It is recommended that each mouse ICAM-1 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty