Would you like to log in again?
Human CD30 - TNFRSF8 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0346
76237-598EA 636 USD InStock
76237-598
Human CD30 - TNFRSF8 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human CD30 - TNFRSF8 AccuSignal ELISA Kit
  • Natural and recombinant human CD30.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: F19-K379
CD30, also known as TNFRSF8, is a cell membrane protein of the tumor necrosis factor receptor family and tumor marker. This receptor is a positive regulator of apoptosis, and also has been shown to limit the proliferative potential of autoreactive CD8 effector T cells and protect the body against autoimmunity. This gene is mapped to 1p36.22. CD30 is expressed in embryonal carcinoma but not in seminoma and is thus a useful marker in distinguishing between these germ cell tumors. CD30 mast cell activation represents an IgE-independent activation pathway, which is important for understanding cutaneous inflammation associated with mast cells. In addition to those, CD30 is also associated with anaplastic large cell lymphoma.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 313pg/ml, 156pg/ml human CD30 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma (heparin, EDTA) to each empty well. It is recommended that each human CD30 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty