Would you like to log in again?
Mouse CD30L ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0348
76237-602EA 652 USD InStock
76237-602
Mouse CD30L ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse CD30L AccuSignal ELISA Kit
  • Natural and recombinant mouse CD30L.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: Q68-D239
The human CD30L(CD153) gene is located at 9q33. It contains 4 exons and spans more than 17.1 kb. CD153 is expressed on the surface of B cells and this expression is upregulated upon CD154(CD40LG), IL4, and B-cell receptor engagement. In these cells, engagement of CD153 by T cell CD30 inhibits immunoglobulin class switch recognition as well as IgG, IgA, and IgE production, suggesting that this"reverse signaling" modulates the CD154-dependent switching of B cells into the pool producing IgG, IgA, and IgE. Additionally, recombinant human CD30L enhanced the proliferation of CD3 -activated T cells, but induced differential responses, including cell death, in several CD30-positive lymphoma-derived cell lines.The standard product used in this kit is recombinant mouse CD30L with the molecular mass of 30-45KDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.3pg/ml, 15.6pg/ml mouse CD30L standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. We recommend that each mouse CD30L standard solution and each sample is measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty