Would you like to log in again?
Human KLK5 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0487
76237-614EA 687 USD InStock
76237-614
Human KLK5 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human KLK5 AccuSignal ELISA Kit
  • Natural and recombinant human Kallikrein 5.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: V23-S293
Kallikrein gene 5(KLK5, also known as KLK-L2), located on chromosome 19q13.4, is one of the newly identified members of the kallikrein gene family, which is a subgroup of the serine protease enzyme family. In normal human tissues, KLK5 is highly expressed in skin, mammary gland and testis. KLK5 has been suggested to regulate cell shedding(desquamation) in conjunction with KLK7 and KLK14, given its ability to degrade proteins which form the extracellular component of cell junctions in the stratum corneum. It is proposed that KLK5 regulates this process since it is able to self-activate in addition to activating KLK7 and KLK14.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml, 78pg/ml human KLK5 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma(heparin, EDTA), human milk or saliva to each empty well. We recommend that each human KLK5 standard solution and each sample is measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty