Would you like to log in again?
Mouse Chordin ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KOA0845
76237-638EA 636 USD InStock
76237-638
Mouse Chordin ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse Chordin AccuSignal ELISA Kit
  • Natural and recombinant mouse Chordin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: T27-S948
Chordin is a key developmental protein that dorsalizes early vertebrate embryonic tissues by binding to ventralizing TGF-beta-like bone morphogenetic proteins (BMPs) and sequestering them in latent complexes. In mice, Chordin is expressed in the node but not in the anterior visceral endoderm. It has been found to be required for forebrain development. In developing mice that are deficient in both chordin and noggin, the head is nearly absent. This is significant because when only noggin is deficient there are mild defects but the head still forms. Chordin is also involved in avian gastrulation. It is expressed in the anterior cells of Koller's sickle, which form the anterior cells of the primitive streak, a key structure through which gastrulation occurs.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml mouse Chordin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. We recommend that each mouse Chordin standard solution and each sample is measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty