Would you like to log in again?
Human Stanniocalcin 1 - STC1 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0847
76237-642EA 636 USD InStock
76237-642
Human Stanniocalcin 1 - STC1 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Stanniocalcin 1 - STC1 AccuSignal ELISA Kit
  • Natural and recombinant human STC1.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: V34-A247
Stanniocalcin-1 is a glycoprotein which is encoded by the STC1 gene. This gene encodes a secreted, homodimeric glycoprotein that is expressed in a wide variety of tissues and may have autocrine or paracrine functions. Human Stanniocalcin-1 is a putative molecular biomarker of leukemic microenvironment and the only molecular function known up to date is a SUMO E3 ligase activity in the SUMOylation cycle. STC1 interacts with lots of proteins in the cytoplasm, mitochondria, endoplasmatic reticulum and dot-like fashion in the nucleus. The N-terminal region of STC1 is the function region which is responsible to establish the interaction with its partners, including SUMO1. Low resolution studies shows that STC1 is an anti-parallel homodimer in solution and the cystein 202 is responsible for the dimerization of this protein. All the 5 dissulfide bonds of human STC1 are conserved and have the same profile of fish STC.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml human STC1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum and plasma (heparin, EDTA) to each empty well. It is recommended that each human STC1 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty