Would you like to log in again?
Packing Sheets, Extra Packaging Corporation
Supplier: EXTRA PACKAGING CORP. TE
20-100-6
76237-916EA 991.64 USD InStock
76237-916
Packing Sheets, Extra Packaging Corporation
Seals General Purpose Sealing Films
Anti-static, fire retardant film that protects surfaces during periods of installation, construction, maintenance, repair, tenant improvement, hospital renovation and telecommunication work.
  • Printed with FR standard and a statement
  • Can be used to cover a variety of surfaces including electronic equipment
  • Anti-Static nature of this film does not attract dust from its environment
A perfect protective covering for equipment sensitive to electrostatic discharge.
Certifications: Passes the NFPA 701-04 Test 1 Fire Standard and ASTM E-84 Class A.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty