Would you like to log in again?
Tin Can with One-Touch Pump, Menda
Supplier: Menda
35335
76238-038EA 25.12 USD InStock
76238-038
Tin Can with One-Touch Pump, Menda
Bottles Dropping and Dispensing Bottles
Tin construction allows storage of fluid not compatible with plastic and most commonly used when strength and durability are important.
  • Ideal for use with hand sanitizer
One handed operation amkes this as perfect for cleaning with swabs and brushes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty