Would you like to log in again?
3M™ Quat Disinfectant Cleaner 5H, Dalco
Supplier: Dalco
3M™
23218
76238-050CS 1025.73 USD InStock
76238-050
3M™ Quat Disinfectant Cleaner 5H, Dalco
Cleaning Agents Disinfectants
3M™ Quat 5H Disinfectant Cleaner is an EPA-registered, hospital-grade quaternary disinfectant cleaner.
  • Rinse-free, pleasant fragrance
  • High flow for dispensing into buckets
Kills HIV-1, MRSA, VRE, Herpes Simplex I and II and other pathogens.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty