Would you like to log in again?
Clamp Gaskets, EDPM, Garlock Hygienic Technologies
Supplier: Garlock
40MPE-400 40MPE-100
76238-126EA 1.71 USD InStock
76238-126 76238-128
Clamp Gaskets, EDPM, Garlock Hygienic Technologies
Seals Gaskets
Tri-clamp sanitary gaskets for use in pharmaceutical industries as well as in Food and Beverage.
  • Blue gasket is X-ray detectable
  • For use with 1 inch (black) or 4 inch (blue) tubes
Tri-clamp sanitary gaskets fit into the grooves of a tri-clamp tube fitting. Tri-clamp fittings, also called tri-clover fittings, clamp together two tubes in a process line and the gasket provides a sterile, leakproof seal.
Certifications: Meets 21CFR 177.2600; passed U.S.P. Class VI Cytotoxicity testing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty