Would you like to log in again?
Sanitary Tri-Clamp® Standard Gaskets, Garlock Hygienic Technologies
Supplier: Garlock
40MPU-150 40MPG-100 40MPG-150
76238-134EA 2.28 USD InStock
76238-134 76238-142 76238-132
Sanitary Tri-Clamp® Standard Gaskets, Garlock Hygienic Technologies
Seals Gaskets
Sanitary gaskets are used widely in the food, dairy, beverage, biotech, pharmaceutical, and all other sanitary process industries to seal clamp connections in sanitary pipe lines.
Rubber fab sanitary Tri-Clamp® gaskets are ADI free (Animal Derived Ingredient free). Buna N is not USP class VI or ADI free.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty