Would you like to log in again?
Kwik Clamp™ Double Bond Hose Clamps, Newage Industries
Supplier: NewAge Industries
570-0457 570-0996 570-0611
76238-254EA 1.34 USD InStock
76238-254 76238-256 76238-258
Kwik Clamp™ Double Bond Hose Clamps, Newage Industries
Clamps Tubing and Sanitary Clamps
Kwik Clamp™ are typically stored and shipped with a small amount of water. This helps to maintain physical properties and prevents the clamps from drying and/or cracking prior to use. Water does not harm the product.
  • Easy to install and remove with hose in place
  • Reusable
  • Heat and light stabilized
  • Will not rust or corrode
  • Patented double row of teeth ensures a strong bond that will not vibrate loose
Certifications: RoHS compliant.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty